missing translation for 'onlineSavingsMsg'
Learn More

Diflunisal Phosphate, TRC

Product Code. 30388142
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388142 100 mg 100mg
30372614 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388142 Supplier Toronto Research Chemicals Supplier No. TRCD445750100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diflunisal Phosphate
CAS 84958-45-2
Molecular Formula C13 H9 F2 O6 P
Quantity 100 mg
InChI Formula InChI=1S/C13H9F2O6P/c14-8-2-3-9(11(15)6-8)7-1-4-12(21-22(18,19)20)10(5-7)13(16)17/h1-6H,(H,16,17)(H2,18,19,20)
SMILES OC(=O)c1cc(ccc1OP(=O)(O)O)c2ccc(F)cc2F
IUPAC Name 5-(2,4-difluorophenyl)-2-phosphonooxybenzoic acid
Molecular Weight (g/mol) 330.18
Formula Weight 330.0105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.