missing translation for 'onlineSavingsMsg'
Learn More

Diflunisal, MedChemExpress

Product Code. 16417466
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417466 Supplier Medchem Express Supplier No. HY18342
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Diflunisal (MK-647) is a salicylate derivative with nonsteroidal anti-inflammatory and uricosuric properties, which is used alone as an analgesic and in rheumatoid arthritis patients. The mechanism of action of diflunisal is as a Cyclooxygenase (COX) Inhibitor.

Diflunisal (MK-647) is a salicylate derivative with nonsteroidal anti-inflammatory and uricosuric properties, which is used alone as an analgesic and in rheumatoid arthritis patients. The mechanism of action of diflunisal is as a Cyclooxygenase (COX) Inhibitor.

Specifications

Chemical Name or Material Diflunisal
CAS 22494-42-4
Color White
Purity Grade Notes Research
Molecular Formula C13H8F2O3
Quantity 100 mg
Synonym MK-647
Solubility Information DMSO : 50 mg/mL (199.84 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=CC=C(F)C=C2F)=CC=C1O)O
Molecular Weight (g/mol) 250.2
Formula Weight 250.2
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.