missing translation for 'onlineSavingsMsg'
Learn More

Diflubenzuron, TRC

Product Code. 30134801
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134801 1 g 1g
30138536 10 g 10g
17900345 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134801 Supplier Toronto Research Chemicals Supplier No. TRCD4455201G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35367-38-5
Molecular Formula C14 H9 Cl F2 N2 O2
Molecular Weight (g/mol) 310.68
Synonym N-[[(4-Chlorophenyl)amino]carbonyl]-2,6-difluorobenzamide, 1-(4-Chlorophenyl)-3-(2,6-difluorobenzoyl)urea, AI 3-29054, Diflubenzuron, Difluron, Dimilin, Dimilin 25, Du-Dim, Duphar PH 60-40, Larvakil, Micromite, Micromite (Uniroyal), Minerin, Mosdop P, Mosdop TB, N-(2,6-Difluorobenzoyl)-N'-(4-chlorophenyl)urea, N-(p-Chlorophenyl)-N'-(2,6-difluorobenzoyl)urea, OMS 1804, PH 60-40, Rotadin, Sniper, Sniper (benzoylurea insecticide), Suniao-one, TH 6040, Thompson-Hayward 6040
IUPAC Name N-[(4-chlorophenyl)carbamoyl]-2,6-difluorobenzamide
SMILES Fc1cccc(F)c1C(=O)NC(=O)Nc2ccc(Cl)cc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H9ClF2N2O2/c15-8-4-6-9(7-5-8)18-14(21)19-13(20)12-10(16)2-1-3-11(12)17/h1-7H,(H2,18,19,20,21)
Formula Weight 310.0321
Chemical Name or Material Diflubenzuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.