missing translation for 'onlineSavingsMsg'
Learn More

Diflubenzuron-d4 (4-chlorophenyl-d4), TRC

Product Code. 30382664
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382664 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382664 Supplier Toronto Research Chemicals Supplier No. TRCD4455215MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diflubenzuron chlorophenyl-d4
CAS 1219795-45-5
Molecular Formula C14 D4 H5 Cl F2 N2 O2
Quantity 5 mg
Synonym Diflubenzuron D4 (Chlorophenyl D4), 1-(4-Chlorophenyl)-3-(2,6-difluorobenzoyl)urea D4, Benzamide, N-[[(4-chlorophenyl)amino]- carbonyl]-2,6-difluoro- D4, Diflubenzuron D4
InChI Formula InChI=1S/C14H9ClF2N2O2/c15-8-4-6-9(7-5-8)18-14(21)19-13(20)12-10(16)2-1-3-11(12)17/h1-7H,(H2,18,19,20,21)/i4D,5D,6D,7D
SMILES [2H]c1c([2H])c(NC(=O)NC(=O)c2c(F)cccc2F)c([2H])c([2H])c1Cl
IUPAC Name N-[(4-chloro-2,3,5,6-tetradeuteriophenyl)carbamoyl]-2,6-difluorobenzamide
Molecular Weight (g/mol) 314.7079
Formula Weight 314.0572
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.