missing translation for 'onlineSavingsMsg'
Learn More

Difloxacin-d3 (methyl-d3), TRC

Product Code. 30383714
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383714 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30383714 Supplier Toronto Research Chemicals Supplier No. TRCD4456195MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Difloxacin-d3 (methyl-d3)
CAS 1173147-93-7
Molecular Formula C21 D3 H16 F2 N3 O3
Quantity 5 mg
Synonym 6-Fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(trideuteriomethyl)piperazin-1-yl]quinoline-3-carboxylic acid, 6-Fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(methyl-d3)piperazin-1-yl]quinoline-3-carboxylic acid, Difloxacin-D3 (N-methyl-D3), Difloxacin-d3
InChI Formula InChI=1S/C21H19F2N3O3/c1-24-6-8-25(9-7-24)19-11-18-15(10-17(19)23)20(27)16(21(28)29)12-26(18)14-4-2-13(22)3-5-14/h2-5,10-12H,6-9H2,1H3,(H,28,29)/i1D3
SMILES [2H]C([2H])([2H])N1CCN(CC1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)c4ccc(F)cc4
IUPAC Name 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(trideuteriomethyl)piperazin-1-yl]quinoline-3-carboxylic acid
Molecular Weight (g/mol) 402.4092
Formula Weight 402.1583
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.