missing translation for 'onlineSavingsMsg'
Learn More

Diflorasone, MedChemExpress

Product Code. 16483776
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483776 25 mg 25mg
16480738 5 mg 5mg
16478036 100 mg 100mg
16448308 10 mg 10mg
16404197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483776 Supplier Medchem Express Supplier No. HYA0158
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diflorasone act as a corticosteroid hormone receptor agonist with anti-inflammatory and immunosuppressive properties. Diflorasone enters the cell by diffusion across the cell membrane and binds to the glucocorticoid receptor (GR) in the cytoplasm. Diflorasone is used for the research of skin diseases such as eczema or psoriasis.

Diflorasone act as a corticosteroid hormone receptor agonist with anti-inflammatory and immunosuppressive properties. Diflorasone enters the cell by diffusion across the cell membrane and binds to the glucocorticoid receptor (GR) in the cytoplasm. Diflorasone is used for the research of skin diseases such as eczema or psoriasis.

Specifications

Chemical Name or Material Diflorasone
CAS 2557-49-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H28F2O5
Quantity 25 mg
Solubility Information DMSO : 130 mg/mL (316.73 mM; Need ultrasonic)
SMILES C[C@@]12[C@](C(CO)=O)(O)[C@@H](C)C[C@@]1([H])[C@]3([H])C[C@H](F)C4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C2)=O
Molecular Weight (g/mol) 410.45
Formula Weight 410.45
Percent Purity 98.96%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.