missing translation for 'onlineSavingsMsg'
Learn More

Difenzoquat bis-Trideuteromethyl Methyl-D3 Perchlorate Salt, TRC

Product Code. 30385537
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385537 10 mg -
30371618 50 mg -
30377118 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30385537 Supplier Toronto Research Chemicals Supplier No. TRCD44541910MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C17H11D6N2 • ClO4
Molecular Weight (g/mol) 354.82
Synonym 1,2-Dimethyl-3,5-diphenyl-1H-pyrazolium Methyl Sulfate-D6, Avenge CP-D6
IUPAC Name 1,2-bis(methyl-d3)-3,5-diphenyl-1H-pyrazol-2-iumPerchlorate Salt
SMILES O=Cl([O])([O-])=O.[2H]C([N+](N1C([2H])([2H])[2H])=C(C2=CC=CC=C2)C=C1C3=CC=CC=C3)([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H17N2.ClHO4/c1-18-16(14-9-5-3-6-10-14)13-17(19(18)2)15-11-7-4-8-12-15;2-1(3,4)5/h3-13H,1-2H3;(H,2,3,4,5)/q+1;/p-1/i1D3,2D3;
Formula Weight 354.13
Chemical Name or Material Difenzoquat bis-Trideuteromethyl Methyl-D3 Perchlorate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.