missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Yeast Extract, ultra-filtered (UF)

Product Code. 16219741
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
This item is not returnable. View return policy

Product Code. 16219741

Brand: Gibco™ 210934

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Difco™ Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast.

  • Difco Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
  • Difco Yeast Extract Low-Dusting (LD) was designed to limit the amount of dust generated compared to other spray-dried yeast extracts
  • It is ultra-filtered and specifically designed for mammalian cell culture applications
  • With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum
  • It has an endotoxin level of less than or equal to 500 EU/g
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance This product does not contains HAPs; This product does not contain any Proposition 65 chemicals; WHMIS Hazard Class - Non-controlled - This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations (CPR)and the SDS contains all the information required by the CPR
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Cell Type Bacteria, Insect, Mammalian
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Mammalian, Microbial, Vaccine, Insect cell culture applications
Product Line Difco
Quantity 10 kg
Source Yeast, animal origin-free
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.