missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Yeast Extract, low-dusting (LD)

Product Code. 16209741 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16209741 500 g 500g
16239741 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16209741 Supplier Gibco™ Supplier No. 210933
Make Your Process Development Count
Use promo code 27978
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulation of culture media for cultivation of heterotrophic microorganisms

  • Yeast extract designed to limit the amount of dust generated compared to other spray-dried yeast extracts
  • Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells
  • Gibco™ yeast extracts are derived from primary grown baker’s yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Cell Type Bacteria
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Product Line Difco
Quantity 500 g
Source Yeast, animal origin-free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.