missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Xylose Lysine Deoxycholate (XLD) Agar
Description
- Moderately selective dehydrated medium
- Isolation and differentiation of enteric pathogens, especially Shigella species
- Certifications: AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP |
| Form | Dehydrated |
| Media Type | XLD Agar |
| Formulation | Xylose 3.5 g/L, L-Lysine 5 g/L, Lactose 7.5 g/L, Saccharose 7.5 g/L, Sodium Chloride 5 g/L, Yeast Extract 3 g/L, Phenol Red 0.08 g/L, Sodium Desoxycholate 2.5 g/L, Ferric Ammonium Citrate 0.8 g/L, Sodium Thiosulfate 6.8 g/L, Agar 13.5 g/L |
| For Use With (Application) | Isolation and Differentiation of Enteric Pathogens |
| Packaging Type | Pail |
| Quantity | 10 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?