missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Trypticase™ Soy Agar
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| Color | Light beige |
| Form | Solid |
| For Use With (Application) | Used with or without Blood for the cultivation and isolation of a variety of organism |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction