missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Supplement VX with Reconstituting Fluid

Product Code. 11768802
Click to view available options
:
Reconstituting Fluid; Quantity: 10 mL; Packaging Type: Vial
This item is not returnable. View return policy

Product Code. 11768802

Brand: BD 233542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BD Difco™ Supplement VX with Reconstituting Fluid contains a sterile concentrate of essential growth factors V and X.

  • Supplement VX with Reconstituting Fluid is used for supplementing media to culture fastidious microorganisms, particularly Neisseria gonorrhoeae and Haemophilus influenzae.
    • Lyophilized concentrate
    • Contains vitamins, amino acids, coenzymes, and dextrose
  • This supplement is recommended for enriching GC Agar media, Proteose No. 3 Agar, and the selective agars for the isolation of pathogenic Neisseria.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
For Use With (Application) Supplementing Media to Culture Fastidious Microorganisms
Packaging Type Vial
Quantity 100 mL
compliance-icons
Product Identifier
  • Vial Supplement VX w/ Fluid 1X100ml
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see supplemental first aid instructions on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, Diadenine sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.