missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Sabouraud Dextrose Agar

Product Code. 10367423
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
Unit Size:
10kg
500g
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
10367423 10 kg Pail 10kg
12790187 2 kg Pail Each
11758103 500 g Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367423 Supplier BD Supplier No. 210930

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rendered more selective for fungi by the addition of antimicrobics. BD™ Difco™ Sabouraud Dextrose Agar is a general-purpose medium used in the cultivation of yeasts, molds and aciduric microorganisms. It is also recommended for the testing of cosmetics and food.

Difco™ Sabouraud Dextrose Agar is a dehydrated culture media used in qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes.

  • The addition of antimicrobics is a modification designed to increase bacterial inhibition.
  • The low pH of approximately 5.6 is favorable for the growth of fungi and slightly inhibitory to contaminating bacteria in clinical specimens.
  • General Chapters 61 and 62 of the USP describe test methods for using Sabouraud Dextrose Agar when performing the microbial enumeration tests and tests for isolating Candida albicans from nonsterile pharmaceutical products.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Color Beige
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Sabouraud Dextrose Agar
Product Type Agar
Formulation Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes
Packaging Type Pail
Quantity 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.