missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Plate Count Agar

Product Code. 11708942
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11708942

Brand: BD 247910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for obtaining microbial plate counts from milk and dairy products, foods, water and other materials of sanitary importance. BD™ Difco™ Plate Count Agar is a dehydrated medium base that meets APHA requirements.

Plate Count Agar used in the enumeration of bacteria in water, wastewater, food and dairy. It is made according to the American Public Health Association (APHA) formulation. It is recommended for obtaining plate counts for milk and other dairy products and may also be used to determine the sanitary quality of foods, water and other materials. Each lot of dehydrated medium base is subjected to the APHA quality control test.

  • Good cultural response to Lactobacillus johnsonii 11506 and Staphylococcus aureus 25923
  • Enzymatic digest of casein provides amino acids and other complex nitrogenous substances
  • Yeast extract primarily supplies the B-complex vitamins
  • Dextrose is an energy source
  • pH (2.35% solution, 25°C): 7.0 ± 0.2
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW, USDA, APHA
Color Beige
Content And Storage 2°C to 30°C
Form Dehydrated Solid
Media Type Plate Count Agar
Product Type Agar
Formulation Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L
For Use With (Application) For obtaining microbial plate counts from Milk and dairy product, foods, Water and other materials of sanitary importance
Packaging Type Pail
Quantity 2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.