missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Muller Kauffmann Tetrathionate Broth Base

Product Code. 11718283
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11718283

Brand: BD 218531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Broth Type Muller Kauffmann Tetrathionate Broth Base
Color White
Form Powder
Media Type Dehydrated
Product Type Microbiology Media
For Use With (Application) For the selective isolation of Salmonella
Quantity 500 g
compliance-icons
Product Identifier
  • Bottle Muller Kauffmann Tet Broth 500G
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Bile, extract
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.