missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Lactose Peptone Broth
Click to view available options
:
Lactose Peptone Broth; Quantity: 500 g
Description
- Lactose Peptone Broth contains peptones, which provide the carbon and nitrogen sources required for good growth of a wide variety of organisms.
- Lactose is provided as a source of fermentable carbohydrate.
- Sodium chloride is present in the medium to provide a suitable osmotic environment.
- Bromcresol purple is used as a colorimetric indicator to show the production of acid from the fermentation of lactose.
Specifications
Specifications
| Form | Dehydrated |
| Media Type | Lactose Peptone Broth |
| Formulation | Pancreatic Digest of Casein 17.0 g/L, Soy Peptone 3.0 g/L, Lactose 10.0 g/L, Sodium Chloride 5.0 g/L, Bromcresol Purple 0.02 g/L |
| pH | 7.4 ±0.2 |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction