missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Heart Infusion Agar
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
In plates, Heart Infusion Agar can be used for primary isolation of organisms from mixed cultures. In agar slant form, it is used primarily for organism cultivation and maintenance rather than for isolation from mixed cultures.
Specifications
Specifications
| Color | Beige |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction