missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Folic AOAC Medium

Product Code. 11798173
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11798173 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11798173 Supplier BD Supplier No. 212169

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

BD Difco™ Folic AOAC Medium for determining folic acid concentration by the microbiological assay technique.

  • Folic AOAC Medium, microbiological assay of folic acid for determining folic acid concentration by the microbiological assay technique.
  • Certifications: ISO, 9000
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type Folic Acid Assay Medium
Formulation Vitamin Assay Casamino Acids 12 g/L, Dextrose 40 g/L, Sodium Citrate 20 g/L, L-Cystine 0.2 g/L, DL-Tryptophan 0.2 g/L, Adenine Sulfate 20 mg/L, Guanine Hydrochloride 20 mg/L, Uracil 20 mg/L, Thiamine Hydrochloride 2 mg/L, Pyridoxine Hydrochloride 4 mg/L, Riboflavin 2 mg/L, Niacin 2 mg/L, Calcium Pantothenate 400 μg/L, p-Aminobenzoic Acid 200 μg/L, Biotin 0.8 μg/L, Dipotassium Phosphate 1 g/L, Monopotassium Phosphate 1 g/L, Magnesium Sulfate 0.4 g/L, Sodium Chloride 20 mg/L, Ferrous Sulfate 20 mg/L, Manganese Sulfate 20 mg/L
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.