missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Tryptose Agar

Product Code. p-4869122
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11779062

Brand: BD 264300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Cultivation of Brucella and a variety of pathogenic microorganisms

Certifications

BAM, CCAM, COMPF

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Tryptose Agar
Product Type Agar
Formulation Tryptose 20g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Cultivation of a wide variety of fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.