missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Hektoen Enteric Agar
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | HE Agar |
| Product Type | Agar |
| Formulation | Proteose Peptone 12g/L, Yeast extract 3g/L, Bile Salts No. 3 9g/L, Lactose 12g/L, Saccharose 12g/L, Salicin 2g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 5g/L, Ferric Ammonium Citrate 1.5g/L, Agar 14g/L, Bromthymol blue 65mg/L, Acid Fuchsin 0.1g/L |
| For Use With (Application) | Isolation and cultivation of gram negative enteric microorganisms, especially Shigella, from a variety of clinical and nonclinical specimens |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Product Identifier
- Hektoen Enteric Agar
- Warning
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- MIXTURE LIST-Contain: bile extract, sodiumpyruvate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?