missing translation for 'onlineSavingsMsg'
Learn More

Diethylzinc, nominally 15% w/w in hexanes, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 11366299
Change view
Click to view available options
Quantity:
100g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366299 100g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11366299 Supplier Thermo Scientific Alfa Aesar Supplier No. 044880.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Specifications

CAS 557-20-0
Density 0.720 g/mL
Flash Point −40°C (−40°F)
Packaging ChemSeal™ bottles
Molecular Formula C4H10Zn
Linear Formula Zn(C2H5)2
MDL Number MFCD00009021
Quantity 100g
UN Number UN3394
Sensitivity Air and moisture sensitive
Synonym diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
Solubility Information Reacts with water.
InChI Key IPSRAFUHLHIWAR-UHFFFAOYSA-N
SMILES [Zn++].C[CH2-].C[CH2-]
IUPAC Name zinc;ethane
Molecular Weight (g/mol) 123.50
PubChem CID 11185
Formula Weight 123.49
Chemical Name or Material Diethylzinc
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.