missing translation for 'onlineSavingsMsg'
Learn More

Diethylzinc, nominally 15% w/w in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles

Product Code. 16645905
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16645905 800 mL 800mL
16655905 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 16645905 Supplier Thermo Scientific Alfa Aesar Supplier No. H37734.9Z
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 557-20-0 | C4H10Zn | 123.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-20-0
Molecular Formula C4H10Zn
Molecular Weight (g/mol) 123.50
MDL Number MFCD00009021
InChI Key HQWPLXHWEZZGKY-UHFFFAOYSA-N
IUPAC Name diethylzinc
SMILES CC[Zn]CC

Specifications

Quantity 800 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.