missing translation for 'onlineSavingsMsg'
Learn More

Diethylzinc, 0.9M (15 wt%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10598123 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10598123 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10598123 Supplier Thermo Scientific Supplier No. 205511001
 Restricted Item
In Stock
Add to basket
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones
Sea freight only: For EU stock allow up to 15 days delivery.
Normativa: Este producto es un intermedio aislado transportado de acuerdo con el Reglamento (CE) 1907/2006 (REACH). Todos los clientes deben completar una declaración antes de la entrega.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Diethylzinc
Color Brown-Gray
Name Note 1M Solution in Hexane
CAS 557-20-0, 64742-49-0
Boiling Point 118.0°C
Melting Point -39.0°C to +28.0°C
Molecular Formula C4H10Zn
Linear Formula (C2H5)2Zn
MDL Number MFCD00009021
Concentration 1M, exact strength on the certificate of analysis
Density 0.7260g/mL
Merck Index 15, 3151
Synonym diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
Solubility Information Solubility in water: reacts.
InChI Key HQWPLXHWEZZGKY-UHFFFAOYSA-N
SMILES CC[Zn]CC
IUPAC Name zinc;ethane
Molecular Weight (g/mol) 123.50
PubChem CID 11185
Formula Weight 123.5
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.726
Viscosity 0.7 mPa.s (20°C)
Quantity 100 mL
Flash Point −40°C
Physical Form Solution
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Diethylzinc, 0.9M solution in hexane
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Substance which, in contact with water, emits flammable gases Category 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H304-May be fatal if swallowed and enters airways.
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.