missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyltoluenediamine, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyltoluenediamine |
| CAS | 68479-98-1 |
| Molecular Formula | C11 H18 N2 |
| Quantity | 50 mg |
| Synonym | 2,4(or 4,6)-Diethyl-6(or 2)-methyl-1,3-benzenediamine, 3,5-Diethyl-(2,4-or 2,6-)toluenediamine, Addolink 1701, Aradur 5200, Araldite HY 5200, Baytec 110, Baytec 505, Baytec Curative W, Baytec E 505, Curative W, Cure EPI-W, Curehard 10, Curing agent W, DETD, DETDA, DETDA 80, Diethyltoluenediamine, E 100, E 200, Epicure DX 6509, Epicure W, Epikote 6509, Epon W, Ethacure 100, Ethacure 100 Plus, Ethacure 100LC, Ethacure 100S, Ethacure 200, Ethacure 89, Ethacure A 100, Hardener DT, Hartcure 10, JER Cure W, JER Cure WA, Lonzacure DETDA, Lonzacure DETDA 80, PT 2712B1, Q 111, Q 111 (amine), SEIKA-S, Trivex Component B, VP-LS 2214, Vantico HY 5200 |
| InChI Formula | InChI=1S/C11H18N2/c1-4-8-7(3)6-10(12)9(5-2)11(8)13/h6H,4-5,12-13H2,1-3H3 |
| SMILES | CCc1c(C)cc(N)c(CC)c1N |
| IUPAC Name | 2,4-diethyl-5-methylbenzene-1,3-diamine |
| Molecular Weight (g/mol) | 178.274 |
| Formula Weight | 178.147 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?