missing translation for 'onlineSavingsMsg'
Learn More

Diethyltoluenediamine, TRC

Product Code. 30384920
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384920 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384920 Supplier Toronto Research Chemicals Supplier No. TRCD44230825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyltoluenediamine
CAS 68479-98-1
Molecular Formula C11 H18 N2
Quantity 25 mg
Synonym 2,4(or 4,6)-Diethyl-6(or 2)-methyl-1,3-benzenediamine, 3,5-Diethyl-(2,4-or 2,6-)toluenediamine, Addolink 1701, Aradur 5200, Araldite HY 5200, Baytec 110, Baytec 505, Baytec Curative W, Baytec E 505, Curative W, Cure EPI-W, Curehard 10, Curing agent W, DETD, DETDA, DETDA 80, Diethyltoluenediamine, E 100, E 200, Epicure DX 6509, Epicure W, Epikote 6509, Epon W, Ethacure 100, Ethacure 100 Plus, Ethacure 100LC, Ethacure 100S, Ethacure 200, Ethacure 89, Ethacure A 100, Hardener DT, Hartcure 10, JER Cure W, JER Cure WA, Lonzacure DETDA, Lonzacure DETDA 80, PT 2712B1, Q 111, Q 111 (amine), SEIKA-S, Trivex Component B, VP-LS 2214, Vantico HY 5200
InChI Formula InChI=1S/C11H18N2/c1-4-8-7(3)6-10(12)9(5-2)11(8)13/h6H,4-5,12-13H2,1-3H3
SMILES CCc1c(C)cc(N)c(CC)c1N
IUPAC Name 2,4-diethyl-5-methylbenzene-1,3-diamine
Molecular Weight (g/mol) 178.274
Formula Weight 178.147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.