missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylstilbestrol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
10 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 169°C to 175°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 1 g |
Beilstein | 06, III, 5619 |
Merck Index | 15, 3148 |
Solubility Information | Solubility in water: practically insoluble in water. Other solubilities: soluble in ethylether,ethanol,chloroform,aceton,e,methanol,arachis oil,olive oil |
IUPAC Name | 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol |
Show More |
Safety and Handling
Product Identifier
- Diethylstilbestrol
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1A
- H360D-May damage the unborn child.
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'