missing translation for 'onlineSavingsMsg'
Learn More

Diethylsilane, 96+%

Product Code. 11359175
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359175 5 g 5g
11369175 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359175 Supplier Thermo Scientific Alfa Aesar Supplier No. L16468.06
  • ONLINE EXCLUSIVE PRICE
    £57.68
    £46.65 / 5g
    Save £11.03
    19% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 542-91-6 | C4H10Si | 86.209 g/mol

Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.

Solubility
Miscible with terahydrofuran, diethyl ether, 1,4-dioxane and benzene.

Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, oxygen and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-91-6
Molecular Formula C4H10Si
Molecular Weight (g/mol) 86.209
MDL Number MFCD00039877
InChI Key KZZFGAYUBYCTNX-UHFFFAOYSA-N
Synonym diethylsilane, silane, diethyl, unii-t6ea843e4k, c2h5 2sih2, 3-silapentane, silane, diethyl-,, diethylsilylene radical, diethylsilane, high purity min
PubChem CID 6327565
IUPAC Name diethylsilicon
SMILES CC[Si]CC

Specifications

Boiling Point 56°C
Melting Point -132°C
Quantity 5 g
Flash Point −20°C (−4°F)
Odor Mild
Refractive Index 1.392
Sensitivity Moisture sensitive
Density 0.684
UN Number UN1993
Beilstein 1730902
Merck Index 14,3128
Solubility Information Miscible with terahydrofuran,diethyl ether,1,4-dioxane and benzene.
Formula Weight 88.23
Percent Purity ≥98%
Chemical Name or Material Diethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.