missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphoramidous dichloride, 97%

Product Code. 10003640 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10003640

Brand: Thermo Scientific Acros 310090050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1069-08-5 | C4H10Cl2NP | 174.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1069-08-5
Color Colorless
Density 1.1960g/mL
Boiling Point 179.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 104% (Argentometry)
Packaging Glass bottle
Molecular Formula C4H10Cl2NP
Linear Formula (C2H5)2NPCl2
Refractive Index 1.4960 to 1.4980
MDL Number MFCD00013614
Quantity 5 g
Specific Gravity 1.196
Synonym diethylphosphoramidous dichloride, dichloro diethylamino phosphine, diethylaminodichlorophosphine, et2npcl2, acmc-2098mq, diethylamido dichlorophosphite, diethylamino dichlorophosphine, dichloro diethylamido phosphine, dichlorophosphanyl diethylamine, dichlorophosphorous diethylamide
InChI Key BPEMCEULJQTJMI-UHFFFAOYSA-N
SMILES CCN(CC)P(Cl)Cl
IUPAC Name N-dichlorophosphanyl-N-ethylethanamine
Molecular Weight (g/mol) 174.01
PubChem CID 2733289
Formula Weight 174.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethylphosphoramidous dichloride, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.