missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphonoacetic acid, 98%

Codice prodotto. 10643712 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Cambia vista
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Dimensione della confezione:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging Dimensione della confezione
10643712 25 g Glass bottle 25g
10080451 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10643712 Fornitore Thermo Scientific Acros N. del fornitore 330670250
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Formats
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 3095-95-2 | C6H13O5P | 196.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3095-95-2
Molecular Formula C6H13O5P
Molecular Weight (g/mol) 196.14
MDL Number MFCD00192032
InChI Key DVQMPWOLBFKUMM-UHFFFAOYSA-N
Synonym 2-diethoxyphosphoryl acetic acid, diethylphosphonoacetic acid, diethyl carboxymethylphosphonate, diethoxyphosphorylacetic acid, diethyl phosphonoacetate, diethoxyphosphoryl acetic acid, acetic acid, diethoxyphosphinyl, acmc-20al82, diethylphosphonoethanoic acid, diethylcarboxymethylphosphonate
PubChem CID 36704
IUPAC Name 2-diethoxyphosphorylacetic acid
SMILES CCOP(=O)(CC(=O)O)OCC

Specifica

Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 150.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460
Quantity 25 g
Specific Gravity 1.22
Formula Weight 196.14
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material Diethylphosphonoacetic acid, 98%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Diethylphosphonoacetic acid
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.