missing translation for 'onlineSavingsMsg'
Learn More

Diethylphenylphosphine, 97%, Thermo Scientific Chemicals

Product Code. 11371228
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371228 1 g 1g
11381228 5 g 5g
11391228 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11371228 Supplier Thermo Scientific Alfa Aesar Supplier No. 010180.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Diethylphenylphosphine is used as a catalyst for selective cross-dimerization, carboxyl migration reactions, selective hydrogenation, asymmetric induction by chiral diphosphines in ring contraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1605-53-4
Molecular Formula C10H15P
Molecular Weight (g/mol) 166.20
MDL Number MFCD00015172
InChI Key LVTCZSBUROAWTE-UHFFFAOYSA-N
Synonym diethylphenylphosphine, diethylphenylphosphene, diethyl phenyl phosphane, phosphine, diethylphenyl, diethyl phenyl phosphine, phenyldiethylphosphine, diethylphenyl phosphine, acmc-209dl7
PubChem CID 74149
IUPAC Name diethyl(phenyl)phosphane
SMILES CCP(CC)C1=CC=CC=C1

Specifications

Density 0.954g/mL
Boiling Point 220°C
Flash Point 66° (150°F)
Odor Pungent
Refractive Index 1.5475
Quantity 1 g
UN Number UN3278
Sensitivity Air & Moisture Sensitive
Formula Weight 166.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethylphenylphosphine
Show More Show Less
compliance-icons
Product Identifier
  • Diethylphenylphosphine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.