missing translation for 'onlineSavingsMsg'
Learn More

Diethylgermanium dichloride

Product Code. 11387778
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387778 1 g 1g
11397778 5 g 5g
11307788 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387778 Supplier Thermo Scientific Alfa Aesar Supplier No. 034108.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

As an intermediate | CAS: 13314-52-8 | C4H14Cl2Ge | 205.69 g/mol

As an intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13314-52-8
Molecular Formula C4H14Cl2Ge
Molecular Weight (g/mol) 205.69
MDL Number MFCD00013589
InChI Key FVUWAWRMMUJVGO-UHFFFAOYSA-N
Synonym diethylgermanium dichloride, diethyldichlorogermane, diethyldichlorogermanium, dichloro diethyl germane, germane,dichlorodiethyl, acmc-1c1bs, diethylgermanium chloride, dichlorodiethylgermane
PubChem CID 83334
IUPAC Name dichloro(diethyl)germane
SMILES Cl.Cl.CC[GeH2]CC

Specifications

Melting Point -37°C
Color Colorless
Density 1.372g/mL
Boiling Point 175°C
Flash Point 63° (145°F)
Odor Acrid
Refractive Index 1.4710
Quantity 1 g
UN Number UN3265
Sensitivity Moisture Sensitive
Solubility Information Decomposes in water and NH3; Soluble in organic solvents
Formula Weight 201.64
Physical Form Liquid
Chemical Name or Material Diethylgermanium dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.