missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic Acid, TRC

Product Code. 30370645
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370645 10 g
30370491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370645 Supplier Toronto Research Chemicals Supplier No. TRCD44437010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-43-6
Molecular Formula C14 H23 N3 O10
Molecular Weight (g/mol) 393.35
Synonym Glycine, N,N-bis[2-[bis(carboxymethyl)amino]ethyl]-, N,N-Bis[2-[bis(carboxymethyl)amino]ethyl]glycine, 1,1,4,7,7-Diethylenetriaminepentaacetic acid, 3,6,9-Triazaundecanedioic acid, 3,6,9-tris(carboxymethyl)-, Acetic acid, 2,2',2'',2'''-[[(carboxymethyl)imino]bis(2,1-ethanediylnitrilo)]tetrakis-, Chel 330 acid, Chel DTPA, Chelest 3PA, Clewat DA, Complexon V, DETP, DETPA, DPTA, DTPA, Dabeersen 503, Detapac, Detarex, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid, Diethylenetriaminepentaacetic acid, Dissolvine D, Hamp-Ex Acid, Monaquest CAI, NSC 7340, Pentacarboxymethyl diethylenetriamine, Pentetic acid, Titriplex V, Trilon CS, [[(Carboxymethyl)imino]bis(ethylenenitrilo)]tetraacetic acid
IUPAC Name 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES OC(=O)CN(CCN(CC(=O)O)CC(=O)O)CCN(CC(=O)O)CC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)
Formula Weight 393.1383
Percent Purity >95%
Chemical Name or Material Pentetic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.