missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution

Product Code. p-3735836 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
3 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
3kg
This item is not returnable. View return policy

Product Code. 10116613

Brand: Thermo Scientific Chemicals 407292500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylenetriaminepentaacetic acid, pentasodium salt
Name Note 40% Aqueous Solution
CAS 140-01-2, 7732-18-5
Melting Point -40°C
Density 1.2900g/mL
pH 11.0 to 12.0 (1% aq. soln.)
Boiling Point 106°C
Flash Point >100°C
Packaging Glass bottle
Molecular Formula C14H18N3Na5O10
Linear Formula NaOCOCH2N(CH2CH2N(CH2COONa)2)2
Refractive Index 1.4185 to 1.4205
MDL Number MFCD00051016
Quantity 250 g
Beilstein 04, III, 1190
Specific Gravity 1.29
Synonym pentasodium dtpa, tetralon b, trilon c, versenex 80, pentasodium pentetate, hamp-ex 80, detarex py, kiresuto p, chelest p, plexene d
Solubility Information Solubility in water: soluble. Other solubilities: mixible with many organic solvents
InChI Key LQPLDXQVILYOOL-UHFFFAOYSA-I
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O
IUPAC Name pentasodium;2-[bis[2-[bis(carboxylatomethyl)amino]ethyl]amino]acetate
Molecular Weight (g/mol) 503.26
PubChem CID 8779
Formula Weight 503.26
Percent Purity 39 to 41%
Grade Technical
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.