missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriamine, TRC

Product Code. 30385096
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385096 50 g -
30385957 10 g 10g
30373869 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385096 Supplier Toronto Research Chemicals Supplier No. TRCD44425550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-40-0
Molecular Formula C4H13N3
Molecular Weight (g/mol) 103.17
Synonym N-(2-aminoethyl)-1,2-ethanediamine, 1,4,7-Triazaheptane, 1,5-Diamino-3-azapentane, 2,2'-Diaminodiethylamine, 2,2'-Iminobis(ethanamine), 2-(2-Aminoethylamino)ethylamine, 3-Azapentane-1,5-diamine, 3-Azapentanediyl-1,5-diamine, Ancamine DETA, Bis(2-Aminoethyl)amine, Bis(beta-Aminoethyl)amine, ChS-P 1, D 93856, DEH 20, DETA, Epicure 3223, Epicure 3290, Epicure T, Epon 3223, H 9506, JER Cure T, JER-T, N,N-Bis(2-Aminoethyl)amine, N-(2-Aminoethyl)-1,2-ethanediamine, N-(2-Aminoethyl)ethylenediamine, NCI 138881, NSC 446, P 11, P 11 (Crosslinking Agent)
IUPAC Name N'-(2-aminoethyl)ethane-1,2-diamine
SMILES NCCNCCN

Specifications

Quantity 50 g
InChI Formula InChI=1S/C4H13N3/c5-1-3-7-4-2-6/h7H,1-6H2
Formula Weight 103.1109
Chemical Name or Material Diethylenetriamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.