missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol monomethyl ether, 98%

Product Code. 11433077
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433077 2500 mL 2500mL
11423077 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433077 Supplier Thermo Scientific Alfa Aesar Supplier No. A16063.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 111-77-3 | C5H12O3 | 120.15 g/mol

Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00002871,MFCD00084416
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
Synonym 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
PubChem CID 8134
ChEBI CHEBI:44836
IUPAC Name 2-(2-methoxyethoxy)ethanol
SMILES COCCOCCO

Specifications

Melting Point -70°C
Density 1.01
Quantity 2500 mL
Flash Point 83°C (181°F)
Odor Light
Refractive Index 1.426
Linear Formula CH3OCH2CH2OCH2CH2OH
Beilstein 1697812
Merck Index 14,6037
Boiling Point 194°C
Solubility Information Fully miscible in water.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material Diethylene glycol monomethyl ether, stabilized with 50 to 150mg/kg BHT
Show More Show Less
compliance-icons
Product Identifier
  • Diethylene glycol monomethyl ether, stab. with 50-150mg
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Diethylene glycol monomethyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.