missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monoethyl Ether, ReagentPlus™ 99%, Honeywell™

Product Code. 15685780 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
Packaging:
Glass bottle
Stainless steel drum
Unit Size:
1 L
100mL
20 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15685780 20 L Stainless steel drum 20 L
15665780 100 mL Glass bottle 100mL
15675780 1 L Glass bottle 1 L
15668420 4 L Glass bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15685780 Supplier Honeywell Research Chemicals Supplier No. 53761620L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-90-0
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

Density 0.999g/mL at 25°C
Packaging Stainless steel drum
Linear Formula C2H5OCH2CH2OCH2CH2OH
Refractive Index 1.427
Quantity 20 L
UN Number NONH for all modes of transport
Beilstein 1736441
Vapor Density 4.63 (vs air)
Formula Weight 134.17g/mol
Percent Purity ≥99.9%
Chemical Name or Material Diethylene glycol monoethyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.