missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Dimethyl Ether(Diglyme), TRC

Product Code. 30371735
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30371735 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371735 Supplier Toronto Research Chemicals Supplier No. TRCD44630010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-96-6
Molecular Formula C6 H14 O3
Quantity 10 g
Synonym Ether, bis(2-methoxyethyl) (6CI,8CI), 1,1'-Oxybis[2-methoxyethane], (2-Methoxyethyl) ether, 1,5-Dimethoxy-3-oxapentane, 1-(2-Methoxyethoxy)-2-methoxyethane, 2,5,8-Trioxanonane, 2-(2-Methoxyethoxy)-1-methoxyethane, Ansul Ether 141, B 0498, Bis(2-methoxyethyl) ether, DMDG, Di(2-methoxyethyl) ether, Diethylene glycol dimethyl ether, Diglyme, Dimethyl carbitol, Dimethyl digol, Ethanol, 2,2'-oxybis-, dimethyl ether, Glyme 2, Glyme 3, Hisolve MDM, Methyl diglyme, NSC 59726, Poly-Solv
InChI Formula InChI=1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3
SMILES COCCOCCOC
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
Molecular Weight (g/mol) 134.17
Formula Weight 134.0943
Chemical Name or Material Bis(2-methoxyethyl)ether (Diethylene glycol dimethyl ether)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.