missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dimethyl ether, 99%, stab. with 100ppm BHT

Product Code. 11465403
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465403 500 mL 500mL
11475403 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11465403 Supplier Thermo Scientific Alfa Aesar Supplier No. A13397.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol

Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with ether.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-96-6
Molecular Formula C6H14O3
MDL Number MFCD00008503
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
Synonym diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
PubChem CID 8150
ChEBI CHEBI:46784
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
SMILES COCCOCCOC

Specifications

Melting Point -64°C
Density 0.944
Boiling Point 161°C to 162°C
Flash Point 51°C (124°F)
Odor Ether-like
Linear Formula (CH3OCH2CH2)2O
Refractive Index 1.408
Quantity 500 mL
UN Number UN3271
Beilstein 1736101
Sensitivity Hygroscopic
Merck Index 14,3165
Solubility Information Miscible with water,ethanol,acetone,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with ether.
Molecular Weight (g/mol) 134.175
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material Diethylene glycol dimethyl ether, stabilized with 100ppm BHT
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethylene glycol dimethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H360FD-May damage fertility. May damage the unborn child.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Diethylene glycol dimethyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.