missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dimethyl ether, Spectrophotometric Grade, ≥99%, Honeywell Riedel-de Haën™

Product Code. 15612770
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15612770 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15612770 Supplier Honeywell-Riedel-de Haen Supplier No. 2563901L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 111-96-6
CAS Max % 100.0000%
Melting Point -64°C (literature)
Density 0.943 g/mL at 25 °C(lit.)
Boiling Point 162°C
Flash Point 57°C
Packaging Glass Bottle
Molecular Formula C6H14O3
Linear Formula C2H5OCH2CH2OH
MDL Number MFCD00002869
Quantity 1 L
UN Number UN3271
Beilstein 1736101
Synonym diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
SMILES COCCOCCOC
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
Molecular Weight (g/mol) 134.175
PubChem CID 8150
Vapor Density 4.6 (vs air)
ChEBI CHEBI:46784
Formula Weight 134.17
Percent Purity >99%
Grade Sprectrophotometric
Chemical Name or Material Diethylene glycol dimethyl ether, Spectrophotometric Grade, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.