missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Dimethyl Ether, 99%, Reagent Grade, Honeywell™

Product Code. p-7120384 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Unit Size:
1 L
2.5 L
20 L
500mL
5mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15668490

Brand: Honeywell Research Chemicals M141025ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-96-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00008503
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
Synonym diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
PubChem CID 8150
ChEBI CHEBI:46784
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
SMILES COCCOCCOC

Specifications

Melting Point -64°C
Color Colorless
Density 0.944g/mL at 20°C, 0.939g/mL at 25°C
Boiling Point 162°C
Flash Point 57°C (134.6°F)
Odor Mild ether-like
Packaging Glass bottle
Linear Formula (CH3OCH2CH2)2O
Quantity 5 mL
UN Number UN3271
Beilstein 1736101
Solubility Information Soluble in water at 20°C
Vapor Density 4.6 (vs air)
Formula Weight 134.17g/mol
Percent Purity 0.99
Chemical Name or Material Diethylene glycol dimethyl ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.