missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Dimethacrylate-d8, TRC
Specifications
Specifications
| Chemical Name or Material | Diethylene Glycol Dimethacrylate-d8 |
| Molecular Formula | C12 D8 H10 O5 |
| Quantity | 10 mg |
| Synonym | 2-Methyl-2-propenoic Acid 1,1'-(Oxydi-2,1-ethanediyl) Ester-d8, Methacrylic Acid Oxydiethylene Ester-d8, Diethylene Glycol Dimethacrylate-d8, 2,2'-Oxybisethanol Dimethacrylate-d8, 2,2'-Oxydiethyl Dimethacrylate-d8, 2G-d8, Blemmer PDE 100-d8, DGM 2-d8, Diethylene Glycol Bis(methacrylate)-d8, Light Ester 2EG-d8, NK Ester 2G-d8, Oxydiethylene Methacrylate-d8, PDE 100-d8, SR 231-d8, SR 231E-d8, TGM 2-d8 |
| InChI Formula | InChI=1S/C12H18O5/c1-9(2)11(13)16-7-5-15-6-8-17-12(14)10(3)4/h1,3,5-8H2,2,4H3/i5D2,6D2,7D2,8D2 |
| SMILES | [2H]C([2H])(OC(=O)C(=C)C)C([2H])([2H])OC([2H])([2H])C([2H])([2H])OC(=O)C(=C)C |
| IUPAC Name | [1,1,2,2-tetradeuterio-2-[1,1,2,2-tetradeuterio-2-(2-methylprop-2-enoyloxy)ethoxy]ethyl] 2-methylprop-2-enoate |
| Molecular Weight (g/mol) | 250.318 |
| Formula Weight | 250.166 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?