missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Diethyl Ether, ≥98%, Reagent Grade, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Glass bottle
Poly bottle
Stainless steel can
Unit Size:
1 L
20 L
4 L
500mL
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | -44°C |
| Color | Colorless |
| Density | 0.909g/mL at 25°C |
| Boiling Point | 189°C (1013 hPa) |
| Flash Point | 67°C (152.6°F) |
| Odor | Weak |
| Packaging | Stainless steel can |
| Linear Formula | (C2H5OCH2CH2)2O |
| Quantity | 20 L |
| Beilstein | 1699259 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction