missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Diethyl Ether, ≥98%, Reagent Grade, Honeywell™

Product Code. p-7120382 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Glass bottle
Poly bottle
Stainless steel can
Unit Size:
1 L
20 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 15617850

Brand: Honeywell Research Chemicals E4658500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-36-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009254
InChI Key RRQYJINTUHWNHW-UHFFFAOYSA-N
Synonym diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
PubChem CID 8179
ChEBI CHEBI:44664
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
SMILES CCOCCOCCOCC

Specifications

Melting Point -44°C
Color Colorless
Density 0.909g/mL at 25°C
Boiling Point 189°C (1013 hPa)
Flash Point 67°C (152.6°F)
Odor Weak
Packaging Glass bottle
Linear Formula (C2H5OCH2CH2)2O
Quantity 500 mL
Beilstein 1699259
Solubility Information Soluble in water at 20°C
Vapor Density 5.6 (vs air)
Formula Weight 162.23g/mol
Percent Purity ≥98%
Chemical Name or Material Diethylene glycol diethyl ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.