missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanophosphonate, 93%, Thermo Scientific™

Product Code. 10167793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167793 100 g Glass bottle 100g
10742651 5 g Glass bottle 5g
10581471 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10167793 Supplier Thermo Scientific Acros Supplier No. 224291000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2942-58-7
Molecular Formula C5H10NO3P
Molecular Weight (g/mol) 163.11
MDL Number MFCD00010256
InChI Key ZWWWLCMDTZFSOO-UHFFFAOYSA-N
Synonym diethyl cyanophosphonate, diethyl phosphorocyanidate, diethylphosphorocyanidate, diethylcyanophosphate, diethyl cyanidophosphate, phosphorocyanidic acid, diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanophosphosphonate, cyanophosphonic acid diethyl ester, diethylcyanophosphonate
PubChem CID 76257
SMILES CCOP(=O)(OCC)C#N

Specifications

Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 104.0°C to 105.0°C (19.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CN
Refractive Index 1.4020 to 1.4040
Quantity 100 g
Fieser 05,17; 06,192; 07,107; 10,145; 15,26; 16,24
Specific Gravity 1.07
Solubility Information Solubility in water: decomposes in water
IUPAC Name diethyl cyanophosphonate
Formula Weight 163.11
Percent Purity 93%
Physical Form Liquid
Chemical Name or Material Diethyl cyanophosphonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.