missing translation for 'onlineSavingsMsg'
Learn More

Diethyl bis(hydroxymethyl)malonate, 95%

Product Code. 10036353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10036353 500 g Glass Bottle 500g
10427760 25 g Glass Bottle 25g
10266770 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10036353 Supplier Thermo Scientific Acros Supplier No. 189535000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20605-01-0 | C9H16O6 | 220.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20605-01-0
Molecular Formula C9H16O6
Molecular Weight (g/mol) 220.22
MDL Number MFCD00009130
InChI Key WIOHBOKEUIHYIC-UHFFFAOYSA-N
Synonym diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h
PubChem CID 311844
IUPAC Name diethyl 2,2-bis(hydroxymethyl)propanedioate
SMILES CCOC(=O)C(CO)(CO)C(=O)OCC

Specifications

Melting Point 45°C to 52°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Linear Formula (HOCH2)2C(CO2C2H5)2
Quantity 500 g
Formula Weight 220.22
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material Diethyl bis(hydroxymethyl)malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.