missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals

Product Code. 10697252
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-30-6
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
MDL Number MFCD00009157
InChI Key DVSDDICSXBCMQJ-UHFFFAOYSA-N
Synonym diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
PubChem CID 66197
IUPAC Name diethyl 2-acetylbutanedioate
SMILES CCOC(=O)CC(C(=O)C)C(=O)OCC

Specifications

Melting Point -8°C
Color Yellow
Density 1.0800g/mL
Boiling Point 180°C to 183°C (50.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C2H5O2CCH2CH(COCH3)CO2C2H5
Refractive Index 1.4340 to 1.436
Quantity 25 g
Beilstein 03, 801
Specific Gravity 1.08
Solubility Information Solubility in water: 15g/L (20°C)
Formula Weight 216.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl acetylsuccinate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'