missing translation for 'onlineSavingsMsg'
Learn More

Diethyle2,5-dibromohexanedioate, 97%, Thermo Scientific™

Product Code. 10153164
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10153164 10 g 10g
10596114 1 g 1g
10722757 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10153164 Supplier Thermo Scientific Maybridge Supplier No. BTB13063EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 869-10-3
Molecular Formula C10H16Br2O4
Molecular Weight (g/mol) 360.04
MDL Number MFCD00075379
InChI Key UBCNJHBDCUBIPB-UHFFFAOYNA-N
Synonym diethyl 2,5-dibromoadipate, diethyl meso-2,5-dibromoadipate, 1,6-diethyl 2,5-dibromohexanedioate, 2,5-dibromoadipic acid diethyl ester, hexanedioic acid, 2,5-dibromo-, diethyl ester, diethyl2,5-dibromohexanedioate, 2,5-dibromohexanedioic acid diethyl ester, acmc-1bjoi, diethylmeso-dibromoadipate, diethylmeso-2,5-dibromoadipate
PubChem CID 241588
IUPAC Name diethyl 2,5-dibromohexanedioate
SMILES CCOC(=O)C(Br)CCC(Br)C(=O)OCC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula [-CH2CH(Br)CO2C2H5]2
Quantity 10 g
Formula Weight 360.04
Percent Purity 97%
Chemical Name or Material Diethyl 2, 5-dibromohexanedioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.