missing translation for 'onlineSavingsMsg'
Learn More

Diethyldithiocarbamic Acid Sodium Salt Trihydrate, TRC

Product Code. 30009366
Change view
Click to view available options
Quantity:
100 mg
250 mg
10 g
Unit Size:
100mg
10g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009366 100 mg 100mg
30003989 10 g 10g
30006206 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009366 Supplier Toronto Research Chemicals Supplier No. TRCD444270100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20624-25-3
Molecular Formula C5 H10 N S2 . Na . 3 H2 O
Molecular Weight (g/mol) 225.31
Synonym Carbamic acid, diethyldithio-, sodium salt, trihydrate (8CI), Carbamodithioic acid, diethyl-, sodium salt, trihydrate (9CI), Sodium N,N-diethyldithiocarbamate trihydrate, Sodium diethyldithiocarbamate trihydrate
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
SMILES O.O.O.[Na+].CCN(CC)C(=S)[S-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1
Formula Weight 225.0469
Percent Purity >95%
Chemical Name or Material N,N-Diethyldithiocarbamate sodium salt trihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.