missing translation for 'onlineSavingsMsg'
Learn More

Diethylcarbamazine Citrate, TRC

Product Code. 30380256
Change view
Click to view available options
Quantity:
500 mg
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380256 5 g
30370644 500 mg
30384482 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380256 Supplier Toronto Research Chemicals Supplier No. TRCD4439005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1642-54-2
Molecular Formula C10 H21 N3 O . C6 H8 O7
Molecular Weight (g/mol) 391.42
Synonym 1-Piperazinecarboxamide, N,N-diethyl-4-methyl-, citrate (1:1) (8CI), 1-Diethylcarbamoyl-4-methylpiperazine dihydrogen citrate, 1-Methyl-4-diethylcarbamoylpiperazine citrate, Banocide, Caricide, Caritrol, Dec, Dicarocide, Diethylcarbamazine Acid Citrate, Diethylcarbamazine citrate, Dirocide, Ditrazin citrate, Ditrazine, Ethodryl citrate, Ethylaminoazine citrate, Filaribits, Filazine, Franocide, Franozan, Hetrazan, Longicid, Loxuran, N,N-Diethyl-4-methyl-1-piperazinecarboxamide dihydrogen citrate, NSC 80513
IUPAC Name N,N-diethyl-4-methylpiperazine-1-carboxamide;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)C(=O)N1CCN(C)CC1.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H21N3O.C6H8O7/c1-4-12(5-2)10(14)13-8-6-11(3)7-9-13;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-9H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 391.1955
Percent Purity >95%
Chemical Name or Material Diethylcarbamazine Citrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.