missing translation for 'onlineSavingsMsg'
Learn More

Diethylbenzene, mixture of isomers

Product Code. 15493187
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493187 2500 mL 2500mL
15403197 100 mL 100mL
15413197 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493187 Supplier Thermo Scientific Alfa Aesar Supplier No. A19548.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25340-17-4 | C10H14

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25340-17-4
Molecular Formula C10H14
MDL Number MFCD00792900
Synonym Dowex J

Specifications

Density 0.87
Boiling Point 182°C to 184°C
Flash Point 55°C (131°F)
Refractive Index 1.495
Quantity 2500 mL
UN Number UN2049
Beilstein 1903396
Formula Weight 134.22
Chemical Name or Material Diethylbenzene, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.